The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005732.3 | NP_005723.2 | 10 - 11 | 2041 | 135 |
GenBank mRNA | AF057299.1 | - | 10 - 11 | 1961 | 135 |
AF057300.1 | - | 10 - 11 | 1896 | 135 | |
AK290597.1 | - | 10 - 11 | 1983 | 135 | |
AK307439.1 | - | 10 - 11 | 1716 | 135 | |
BC073850.1 | - | 10 - 11 | 1958 | 135 | |
BC108282.1 | - | 10 - 11 | 1978 | 135 | |
BC136436.1 | - | 10 - 11 | 1933 | 135 | |
U63139.1 | - | 10 - 11 | 2028 | 135 |