The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005732.3 | NP_005723.2 | 19 - 20 | 3440 | 185 |
GenBank mRNA | AF057299.1 | - | 19 - 20 | 3360 | 185 |
AF057300.1 | - | 19 - 20 | 3295 | 185 | |
BC136436.1 | - | 19 - 20 | 3332 | 185 | |
U63139.1 | - | 19 - 20 | 3427 | 185 |