Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNR
Entrez Gene ID
7143
Gene Name
tenascin R
Gene Aliases
TN-R
Chromosome Location
Chr.1: 175320781 - 175743702 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003285.2 NP_003276.3 16 - 17 3678 60
XM_011509949.2 XP_011508251.1 15 - 16 3287 60
XM_017002218.1 XP_016857707.1 17 - 18 4115 60
XM_017002219.1 XP_016857708.1 16 - 17 4043 60
GenBank mRNA AV650210.2 - 1 - 2 173 60
BC129830.1 - 14 - 15 3001 60
X98085.1 - 15 - 16 3240 60
Z67996.1 - 14 - 15 3204 60

View Full Product Details