The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004963.3 | NP_004954.2 | 17 - 18 | 2073 | 100 |
XM_011520631.2 | XP_011518933.1 | 17 - 18 | 1971 | 100 | |
GenBank mRNA | BC136544.1 | - | 17 - 18 | 1982 | 100 |
BC136545.1 | - | 17 - 18 | 1982 | 100 | |
M73489.1 | - | 17 - 18 | 1984 | 100 |