The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004963.3 | NP_004954.2 | 25 - 26 | 3107 | 80 |
XM_011520631.2 | XP_011518933.1 | 25 - 26 | 3005 | 80 | |
GenBank mRNA | BC136544.1 | - | 25 - 26 | 3016 | 80 |
BC136545.1 | - | 25 - 26 | 3016 | 80 | |
M73489.1 | - | 25 - 26 | 3018 | 80 | |
S57551.1 | - | 25 - 26 | 3087 | 80 |