The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004963.3 | NP_004954.2 | 26 - 27 | 3182 | 128 |
XM_011520631.2 | XP_011518933.1 | 26 - 27 | 3080 | 128 | |
GenBank mRNA | BC136544.1 | - | 26 - 27 | 3091 | 128 |
BC136545.1 | - | 26 - 27 | 3091 | 128 | |
M73489.1 | - | 26 - 27 | 3093 | 128 | |
S57551.1 | - | 26 - 27 | 3162 | 128 |