The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282786.1 | NP_001269715.1 | 2 - 3 | 221 | 103 |
NM_020200.6 | NP_064585.1 | 2 - 3 | 221 | 103 | |
GenBank mRNA | AB209792.1 | - | 2 - 3 | 171 | 103 |
AF226056.1 | - | 2 - 3 | 169 | 103 | |
AK021950.1 | - | 2 - 3 | 181 | 103 | |
AK222674.1 | - | 2 - 3 | 180 | 103 | |
AK294130.1 | - | 2 - 3 | 180 | 103 | |
AK304055.1 | - | 2 - 3 | 180 | 103 | |
AK309614.1 | - | 2 - 3 | 210 | 103 | |
BC008662.2 | - | 2 - 3 | 183 | 103 |