Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFXN3
Entrez Gene ID
81855
Gene Name
sideroflexin 3
Gene Aliases
BA108L7.2, SFX3
Chromosome Location
Chr.10: 101031229 - 101041243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030971.3 NP_112233.2 11 - 12 1336 86
GenBank mRNA AK074707.1 - 10 - 11 949 86
AK093011.1 - 2 - 3 788 86
AK299405.1 - 10 - 11 955 86
BC000124.1 - 10 - 11 942 86

View Full Product Details