The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030971.3 | NP_112233.2 | 6 - 7 | 975 | 59 |
GenBank mRNA | AK299405.1 | - | 5 - 6 | 594 | 59 |
BC000124.1 | - | 5 - 6 | 581 | 59 | |
BG775086.1 | - | 4 - 5 | 451 | 59 |