Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELF2
Entrez Gene ID
10659
Gene Name
CUGBP, Elav-like family member 2
Gene Aliases
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location
Chr.10: 10462515 - 11336675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025076.2 NP_001020247.1 6 - 7 831 68
NM_001025077.2 NP_001020248.1 6 - 7 688 68
NM_001083591.1 NP_001077060.1 6 - 7 831 68
NM_001326317.1 NP_001313246.1 8 - 9 690 68
NM_001326318.1 NP_001313247.1 8 - 9 690 68
NM_001326319.1 NP_001313248.1 9 - 10 728 68
NM_001326320.1 NP_001313249.1 9 - 10 784 68
NM_001326321.1 NP_001313250.1 8 - 9 690 68
NM_001326323.1 NP_001313252.1 9 - 10 784 68
NM_001326324.1 NP_001313253.1 7 - 8 667 68
NM_001326325.1 NP_001313254.1 8 - 9 761 68
NM_001326326.1 NP_001313255.1 7 - 8 686 68
NM_001326327.1 NP_001313256.1 7 - 8 686 68
NM_001326328.1 NP_001313257.1 8 - 9 780 68
NM_001326329.1 NP_001313258.1 6 - 7 720 68
NM_001326330.1 NP_001313259.1 6 - 7 720 68
NM_001326331.1 NP_001313260.1 6 - 7 872 68
NM_001326332.1 NP_001313261.1 6 - 7 872 68
NM_001326334.1 NP_001313263.1 7 - 8 1057 68
NM_001326335.1 NP_001313264.1 6 - 7 872 68
NM_001326336.1 NP_001313265.1 6 - 7 872 68
NM_001326337.1 NP_001313266.1 6 - 7 872 68
NM_001326338.1 NP_001313267.1 5 - 6 778 68
NM_001326339.1 NP_001313268.1 5 - 6 778 68
NM_001326340.1 NP_001313269.1 6 - 7 784 68
NM_001326341.1 NP_001313270.1 6 - 7 784 68
NM_001326342.1 NP_001313271.1 6 - 7 784 68
NM_001326343.1 NP_001313272.1 6 - 7 784 68
NM_001326344.1 NP_001313273.1 6 - 7 574 68
NM_001326345.1 NP_001313274.1 6 - 7 652 68
NM_001326347.1 NP_001313276.1 6 - 7 982 68
NM_001326348.1 NP_001313277.1 6 - 7 980 68
NM_001326349.1 NP_001313278.1 6 - 7 980 68
NM_006561.3 NP_006552.3 6 - 7 777 68
XM_005252354.4 XP_005252411.1 6 - 7 867 68
XM_006717373.2 XP_006717436.1 5 - 6 907 68
XM_011519294.2 XP_011517596.1 6 - 7 867 68
XM_011519297.1 XP_011517599.1 6 - 7 990 68
XM_017015542.1 XP_016871031.1 5 - 6 543 68
XM_017015543.1 XP_016871032.1 6 - 7 633 68
XM_017015544.1 XP_016871033.1 6 - 7 633 68
XM_017015545.1 XP_016871034.1 6 - 7 633 68
XM_017015546.1 XP_016871035.1 6 - 7 633 68
XM_017015547.1 XP_016871036.1 6 - 7 867 68
XM_017015548.1 XP_016871037.1 6 - 7 633 68
XM_017015549.1 XP_016871038.1 6 - 7 633 68
XM_017015550.1 XP_016871039.1 6 - 7 1613 68
XM_017015551.1 XP_016871040.1 6 - 7 867 68
XM_017015552.1 XP_016871041.1 6 - 7 990 68
XM_017015553.1 XP_016871042.1 6 - 7 990 68
XM_017015554.1 XP_016871043.1 6 - 7 990 68
XM_017015555.1 XP_016871044.1 5 - 6 543 68
XM_017015556.1 XP_016871045.1 5 - 6 907 68
XM_017015557.1 XP_016871046.1 7 - 8 572 68
XM_017015558.1 XP_016871047.1 5 - 6 559 68
XM_017015559.1 XP_016871048.1 5 - 6 907 68
XM_017015560.1 XP_016871049.1 5 - 6 568 68
XM_017015561.1 XP_016871050.1 5 - 6 895 68
XM_017015562.1 XP_016871051.1 5 - 6 907 68
XM_017015563.1 XP_016871052.1 5 - 6 894 68
GenBank mRNA AF036956.1 - 6 - 7 747 68
AF090693.1 - 6 - 7 663 68
AF090694.1 - 6 - 7 615 68
AF432906.1 - 6 - 7 683 68
AK293164.1 - 6 - 7 724 68
AK295511.1 - 4 - 5 418 68
AK299548.1 - 6 - 7 607 68
AK299992.1 - 6 - 7 764 68
AK314087.1 - 6 - 7 786 68
AK316354.1 - 6 - 7 583 68
BC036391.1 - 6 - 7 749 68
BP362205.1 - 4 - 5 330 68
DA819821.1 - 5 - 6 501 68
U69546.1 - 6 - 7 564 68

View Full Product Details