The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304811.1 | NP_001291740.1 | 6 - 7 | 845 | 120 |
NM_020374.3 | NP_065107.1 | 6 - 7 | 853 | 120 | |
XM_005253716.1 | XP_005253773.1 | 6 - 7 | 853 | 120 | |
XM_006718992.3 | XP_006719055.1 | 6 - 7 | 853 | 120 | |
GenBank mRNA | AJ272205.1 | - | 6 - 7 | 796 | 120 |
AK074479.1 | - | 6 - 7 | 853 | 120 | |
AL834377.1 | - | 6 - 7 | 816 | 120 | |
BC016908.1 | - | 6 - 7 | 796 | 120 |