The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025065.6 | NP_079341.2 | 2 - 3 | 333 | 98 |
GenBank mRNA | AF322053.1 | - | 2 - 3 | 288 | 98 |
AK022537.1 | - | 2 - 3 | 304 | 98 | |
BC016051.1 | - | 2 - 3 | 295 | 98 |