The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025065.6 | NP_079341.2 | 8 - 9 | 139 | |
GenBank mRNA | AA761809.1 | - | 1 - 2 | 279 | 139 |
AK022537.1 | - | 8 - 9 | 139 | ||
AL359584.2 | - | 6 - 7 | 139 | ||
BC016051.1 | - | 8 - 9 | 139 | ||
CR627468.1 | - | 6 - 7 | 139 |