Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOXL4
Entrez Gene ID
84171
Gene Name
lysyl oxidase like 4
Gene Aliases
LOXC
Chromosome Location
Chr.10: 98247686 - 98268250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032211.6 NP_115587.6 2 - 3 430 65
XM_005270216.1 XP_005270273.1 2 - 3 403 65
XM_006718020.1 XP_006718083.1 3 - 4 593 65
GenBank mRNA AF338441.1 - 1 - 2 279 65
AF395336.1 - 1 - 2 279 65
AK172781.1 - 2 - 3 373 65
AY036093.1 - 1 - 2 285 65
BC035470.1 - 2 - 3 430 65

View Full Product Details