The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032211.6 | NP_115587.6 | 7 - 8 | 1257 | 69 |
XM_005270216.1 | XP_005270273.1 | 7 - 8 | 1233 | 69 | |
XM_006718020.1 | XP_006718083.1 | 8 - 9 | 1423 | 69 | |
GenBank mRNA | AF338441.1 | - | 6 - 7 | 1106 | 69 |
AF395336.1 | - | 6 - 7 | 1106 | 69 | |
AK172781.1 | - | 7 - 8 | 1200 | 69 | |
AY036093.1 | - | 6 - 7 | 1112 | 69 | |
BC013153.1 | - | 7 - 8 | 1257 | 69 | |
BC035470.1 | - | 7 - 8 | 1257 | 69 |