The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308445.1 | NP_001295374.1 | 4 - 5 | 682 | 123 |
NM_025146.3 | NP_079422.1 | 4 - 5 | 685 | 123 | |
GenBank mRNA | AK023090.1 | - | 4 - 5 | 600 | 123 |
AK023256.1 | - | 4 - 5 | 635 | 123 | |
AK296456.1 | - | 4 - 5 | 380 | 123 | |
AK307184.1 | - | 5 - 6 | 529 | 123 | |
AK315876.1 | - | 4 - 5 | 487 | 123 | |
AL601126.1 | - | 4 - 5 | 580 | 123 | |
BC012731.2 | - | 4 - 5 | 624 | 123 |