The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134707.1 | NP_001128179.1 | 3 - 4 | 649 | 69 |
NM_007101.3 | NP_009032.2 | 3 - 4 | 808 | 69 | |
XM_017014367.1 | XP_016869856.1 | 3 - 4 | 811 | 69 | |
XM_017014368.1 | XP_016869857.1 | 3 - 4 | 811 | 69 | |
GenBank mRNA | AF047004.1 | - | 3 - 4 | 647 | 69 |
AF047190.1 | - | 3 - 4 | 647 | 69 | |
AF095735.2 | - | 3 - 4 | 773 | 69 | |
AF095736.2 | - | 2 - 3 | 385 | 69 | |
AF095737.2 | - | 3 - 4 | 777 | 69 | |
AF162428.1 | - | 2 - 3 | 545 | 69 | |
AJ223317.1 | - | 3 - 4 | 647 | 69 | |
AK223402.1 | - | 3 - 4 | 695 | 69 | |
AK291211.1 | - | 3 - 4 | 643 | 69 | |
AK298348.1 | - | 3 - 4 | 692 | 69 | |
AK316494.1 | - | 2 - 3 | 447 | 69 | |
BC136363.1 | - | 3 - 4 | 626 | 69 | |
BC136364.1 | - | 3 - 4 | 626 | 69 | |
BC144035.1 | - | 3 - 4 | 644 | 69 |