Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM7
Entrez Gene ID
2917
Gene Name
glutamate metabotropic receptor 7
Gene Aliases
GLUR7, GPRC1G, MGLU7, MGLUR7, PPP1R87
Chromosome Location
Chr.3: 6861097 - 7743032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000844.3 NP_000835.1 6 - 7 1649 93
NM_181874.2 NP_870989.1 6 - 7 1649 93
XM_017006272.1 XP_016861761.1 6 - 7 1115 93
XM_017006273.1 XP_016861762.1 6 - 7 1115 93
GenBank mRNA AB209214.1 - 4 - 5 640 93
AK097817.1 - 2 - 3 240 93
AK312488.1 - 6 - 7 1649 93
BC136458.1 - 6 - 7 1465 93
BC136459.1 - 6 - 7 1465 93
X94552.1 - 6 - 7 1613 93

View Full Product Details