Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR3A1
Entrez Gene ID
4994
Gene Name
olfactory receptor family 3 subfamily A member 1
Gene Aliases
OLFRA03, OR17-40, OR17-82, OR40
Chromosome Location
Chr.17: 3288318 - 3336087 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002550.2 NP_002541.2 1 - 1 62 80
XM_017024695.1 XP_016880184.1 2 - 2 153 80
XM_017024696.1 XP_016880185.1 2 - 2 127 80
GenBank mRNA BC069414.1 - 1 - 1 62 80
BC096187.3 - 1 - 1 62 80
BC096188.3 - 1 - 1 62 80
BC096189.3 - 1 - 1 62 80
BC096190.1 - 1 - 1 62 80

View Full Product Details