Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKDD1B
Entrez Gene ID
728780
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.5: 75611289 - 75671846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276713.1 NP_001263642.1 12 - 13 1549 65
XM_011543623.2 XP_011541925.1 8 - 9 1301 65
XM_017009814.1 XP_016865303.1 11 - 12 1841 65
XM_017009816.1 XP_016865305.1 9 - 10 1643 65
XM_017009817.1 XP_016865306.1 7 - 8 1235 65

View Full Product Details