The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014271.3 | NP_055086.1 | 7 - 8 | 1411 | 120 |
XM_017029240.1 | XP_016884729.1 | 7 - 8 | 1867 | 120 | |
XM_017029241.1 | XP_016884730.1 | 5 - 6 | 5168 | 120 | |
GenBank mRNA | AB102650.1 | - | 6 - 7 | 903 | 120 |
AF181284.1 | - | 7 - 8 | 1534 | 120 | |
AF284435.1 | - | 7 - 8 | 990 | 120 | |
AJ243874.1 | - | 7 - 8 | 1413 | 120 | |
BC126345.1 | - | 6 - 7 | 942 | 120 | |
BC126347.1 | - | 6 - 7 | 940 | 120 | |
KJ534871.1 | - | 6 - 7 | 903 | 120 |