Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
-
Chromosome Location
Chr.19: 1985438 - 2015703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017797.3 NP_060267.2 2 - 3 526 68
XM_005259593.1 XP_005259650.1 2 - 3 291 68
GenBank mRNA AF355797.1 - 2 - 3 526 68
AK225831.1 - 2 - 3 525 68
AY007116.1 - 2 - 3 248 68
BC094820.1 - 2 - 3 260 68

View Full Product Details