Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS37D
Entrez Gene ID
155382
Gene Name
VPS37D, ESCRT-I subunit
Gene Aliases
WBSCR24
Chromosome Location
Chr.7: 73665346 - 73672110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077621.1 NP_001071089.1 2 - 3 424 136
XM_017011779.1 XP_016867268.1 2 - 3 614 136
GenBank mRNA AY081952.1 - 2 - 3 424 136
BC064621.1 - 2 - 3 271 136

View Full Product Details