The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000877.3 | NP_000868.1 | 9 - 10 | 1325 | 166 |
NM_001288706.1 | NP_001275635.1 | 9 - 10 | 1238 | 166 | |
NM_001320978.1 | NP_001307907.1 | 9 - 10 | 1238 | 166 | |
NM_001320980.1 | NP_001307909.1 | 9 - 10 | 1379 | 166 | |
NM_001320981.1 | NP_001307910.1 | 9 - 10 | 1220 | 166 | |
NM_001320982.1 | NP_001307911.1 | 9 - 10 | 1162 | 166 | |
NM_001320983.1 | NP_001307912.1 | 7 - 8 | 1013 | 166 | |
NM_001320984.1 | NP_001307913.1 | 7 - 8 | 890 | 166 | |
NM_001320985.1 | NP_001307914.1 | 7 - 8 | 911 | 166 | |
XM_005263930.4 | XP_005263987.1 | 9 - 10 | 1187 | 166 | |
XM_005263934.4 | XP_005263991.1 | 10 - 11 | 1292 | 166 | |
XM_011511115.2 | XP_011509417.1 | 10 - 11 | 1284 | 166 | |
XM_011511116.1 | XP_011509418.1 | 9 - 10 | 1262 | 166 | |
XM_011511117.1 | XP_011509419.1 | 9 - 10 | 1257 | 166 | |
XM_011511118.2 | XP_011509420.1 | 9 - 10 | 1188 | 166 | |
XM_017003989.1 | XP_016859478.1 | 9 - 10 | 1385 | 166 | |
GenBank mRNA | AK314433.1 | - | 9 - 10 | 1238 | 166 |
BC067506.1 | - | 8 - 9 | 1059 | 166 | |
BC067507.1 | - | 8 - 9 | 1061 | 166 | |
BC067508.1 | - | 8 - 9 | 1061 | 166 | |
BC075062.2 | - | 8 - 9 | 1041 | 166 | |
BC075063.2 | - | 8 - 9 | 1041 | 166 | |
CD013912.1 | - | 7 - 8 | 882 | 166 | |
CD013913.1 | - | 7 - 8 | 903 | 166 | |
CD014140.1 | - | 7 - 8 | 166 | ||
M27492.1 | - | 8 - 9 | 1079 | 166 | |
X16896.1 | - | 9 - 10 | 1305 | 166 |