The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039580.1 | NP_001034669.1 | 1 - 2 | 197 | 131 |
XM_006714306.3 | XP_006714369.1 | 1 - 2 | 345 | 131 | |
XM_017008618.1 | XP_016864107.1 | 1 - 2 | 175 | 131 | |
GenBank mRNA | AK024812.1 | - | 1 - 2 | 197 | 131 |
AK301075.1 | - | 1 - 2 | 64 | 131 | |
AY690636.1 | - | 1 - 2 | 197 | 131 | |
BC128540.1 | - | 1 - 2 | 131 | ||
BC128541.1 | - | 1 - 2 | 131 | ||
BC146864.1 | - | 1 - 2 | 109 | 131 |