Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT2B
Entrez Gene ID
79979
Gene Name
tRNA methyltransferase 2 homolog B
Gene Aliases
CXorf34, dJ341D10.3
Chromosome Location
Chr.X: 100973358 - 101052118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167970.1 NP_001161442.1 11 - 12 1585 87
NM_001167971.1 NP_001161443.1 11 - 12 1566 87
NM_001167972.1 NP_001161444.1 10 - 11 1528 87
NM_024917.5 NP_079193.2 11 - 12 1978 87
XM_005262195.2 XP_005262252.1 12 - 13 1701 87
XM_005262196.2 XP_005262253.1 10 - 11 1496 87
XM_005262201.2 XP_005262258.1 8 - 9 1076 87
XM_006724705.2 XP_006724768.1 11 - 12 1640 87
XM_011531042.2 XP_011529344.1 8 - 9 1194 87
XM_011531044.2 XP_011529346.1 10 - 11 1255 87
XM_017029858.1 XP_016885347.1 11 - 12 1973 87
XM_017029859.1 XP_016885348.1 10 - 11 1288 87
XM_017029860.1 XP_016885349.1 11 - 12 1587 87
XM_017029861.1 XP_016885350.1 10 - 11 1497 87
XM_017029862.1 XP_016885351.1 10 - 11 1838 87
XM_017029863.1 XP_016885352.1 12 - 13 1510 87
XM_017029864.1 XP_016885353.1 10 - 11 1702 87
XM_017029865.1 XP_016885354.1 10 - 11 1330 87
XM_017029866.1 XP_016885355.1 11 - 12 1393 87
XM_017029867.1 XP_016885356.1 11 - 12 1430 87
XM_017029868.1 XP_016885357.1 10 - 11 1303 87
XM_017029869.1 XP_016885358.1 9 - 10 1647 87
XM_017029870.1 XP_016885359.1 9 - 10 1237 87
XM_017029871.1 XP_016885360.1 9 - 10 1219 87
XM_017029872.1 XP_016885361.1 10 - 11 1330 87
XM_017029873.1 XP_016885362.1 10 - 11 1305 87
XM_017029874.1 XP_016885363.1 9 - 10 1647 87
XM_017029875.1 XP_016885364.1 8 - 9 1194 87
GenBank mRNA AK022749.1 - 10 - 11 1341 87
AK315780.1 - 11 - 12 1735 87
AL832849.1 - 11 - 12 1504 87
BC007526.1 - 10 - 11 1484 87
BC008067.2 - 4 - 5 527 87
BC009437.1 - 10 - 11 1471 87
BC020116.1 - 11 - 12 1539 87
BC034272.2 - 11 - 12 1943 87
BX647969.1 - 10 - 11 1525 87

View Full Product Details