Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TRMT2B
- Entrez Gene ID
-
79979
- Gene Name
- tRNA methyltransferase 2 homolog B
- Gene Aliases
-
CXorf34, dJ341D10.3
- Chromosome Location
-
Chr.X: 100973358 - 101052118 on Build GRCh38
View Full Product Details