The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016239.3 | NP_057323.3 | 12 - 13 | 4825 | 67 |
XM_011523918.2 | XP_011522220.1 | 10 - 11 | 5179 | 67 | |
XM_017024714.1 | XP_016880203.1 | 10 - 11 | 5179 | 67 | |
XM_017024715.1 | XP_016880204.1 | 10 - 11 | 5188 | 67 | |
GenBank mRNA | AF144094.1 | - | 12 - 13 | 4825 | 67 |