The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016239.3 | NP_057323.3 | 3 - 4 | 4032 | 66 |
XM_011523918.2 | XP_011522220.1 | 2 - 3 | 4392 | 66 | |
XM_017024714.1 | XP_016880203.1 | 2 - 3 | 4392 | 66 | |
XM_017024715.1 | XP_016880204.1 | 2 - 3 | 4392 | 66 | |
GenBank mRNA | AF144094.1 | - | 3 - 4 | 4032 | 66 |