Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYO15A
Entrez Gene ID
51168
Gene Name
myosin XVA
Gene Aliases
DFNB3, MYO15
Chromosome Location
Chr.17: 18108706 - 18180227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016239.3 NP_057323.3 45 - 46 8564 103
XM_017024714.1 XP_016880203.1 42 - 43 8864 103
XM_017024715.1 XP_016880204.1 43 - 44 8927 103
GenBank mRNA AF144094.1 - 45 - 46 8564 103
AK294036.1 - 3 - 4 247 103

View Full Product Details