Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYO15A
Entrez Gene ID
51168
Gene Name
myosin XVA
Gene Aliases
DFNB3, MYO15
Chromosome Location
Chr.17: 18108706 - 18180227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016239.3 NP_057323.3 59 - 60 10032 76
XM_017024714.1 XP_016880203.1 56 - 57 10332 76
XM_017024715.1 XP_016880204.1 57 - 58 10395 76
GenBank mRNA AF144094.1 - 59 - 60 10032 76
AK294036.1 - 17 - 18 1715 76
AK297179.1 - 10 - 11 959 76
AK297641.1 - 5 - 6 624 76
AK298825.1 - 5 - 6 409 76
AK310232.1 - 3 - 4 257 76
AL713794.1 - 6 - 7 1144 76
BX538062.1 - 3 - 4 878 76

View Full Product Details