The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101800.2 | NP_001095270.1 | 13 - 14 | 1972 | 97 |
NM_016603.3 | NP_057687.2 | 13 - 14 | 1972 | 97 | |
XM_017009549.1 | XP_016865038.1 | 13 - 14 | 1997 | 97 | |
XM_017009550.1 | XP_016865039.1 | 13 - 14 | 1681 | 97 | |
XM_017009551.1 | XP_016865040.1 | 12 - 13 | 1780 | 97 | |
GenBank mRNA | AF251038.1 | - | 13 - 14 | 1891 | 97 |
AK314224.1 | - | 13 - 14 | 1948 | 97 | |
BC048965.1 | - | 13 - 14 | 1944 | 97 | |
BC056887.1 | - | 12 - 13 | 1547 | 97 |