The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101800.2 | NP_001095270.1 | 2 - 3 | 505 | 126 |
NM_016603.3 | NP_057687.2 | 2 - 3 | 505 | 126 | |
XM_005272007.1 | XP_005272064.1 | 2 - 3 | 505 | 126 | |
XM_006714647.1 | XP_006714710.1 | 2 - 3 | 505 | 126 | |
XM_006714648.1 | XP_006714711.1 | 2 - 3 | 257 | 126 | |
XM_011543448.1 | XP_011541750.1 | 3 - 4 | 298 | 126 | |
XM_011543450.1 | XP_011541752.1 | 2 - 3 | 214 | 126 | |
XM_017009549.1 | XP_016865038.1 | 2 - 3 | 530 | 126 | |
XM_017009550.1 | XP_016865039.1 | 2 - 3 | 214 | 126 | |
GenBank mRNA | AF251038.1 | - | 2 - 3 | 424 | 126 |
AK314224.1 | - | 2 - 3 | 481 | 126 | |
BC048965.1 | - | 2 - 3 | 477 | 126 | |
BC056887.1 | - | 1 - 2 | 80 | 126 | |
BU183000.1 | - | 1 - 2 | 79 | 126 | |
DA499233.1 | - | 2 - 3 | 505 | 126 |