Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST6GAL2
Entrez Gene ID
84620
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 2
Gene Aliases
SIAT2, ST6GalII
Chromosome Location
Chr.2: 106801600 - 106887410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142351.1 NP_001135823.1 2 - 3 1053 81
NM_001142352.1 NP_001135824.1 2 - 3 1123 81
NM_001322362.1 NP_001309291.1 2 - 3 1102 81
NM_032528.2 NP_115917.1 2 - 3 1123 81
XM_006712802.1 XP_006712865.1 2 - 3 1138 81
XM_011511999.2 XP_011510301.1 2 - 3 1235 81
XM_011512000.2 XP_011510302.1 2 - 3 1320 81
XM_011512001.2 XP_011510303.1 2 - 3 1320 81
XM_017005110.1 XP_016860599.1 2 - 3 1074 81
GenBank mRNA AB058780.1 - 2 - 3 1053 81
AB059555.1 - 2 - 3 1109 81
AK307784.1 - 2 - 3 1123 81
BC008680.1 - 2 - 3 1109 81

View Full Product Details