The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291694.1 | NP_001278623.1 | 3 - 4 | 633 | 72 |
NM_003298.4 | NP_003289.2 | 4 - 5 | 690 | 72 | |
XM_011534058.2 | XP_011532360.1 | 7 - 8 | 725 | 72 | |
XM_011534059.2 | XP_011532361.1 | 7 - 8 | 1631 | 72 | |
XM_011534061.2 | XP_011532363.1 | 5 - 6 | 579 | 72 | |
XM_011534063.2 | XP_011532365.1 | 6 - 7 | 666 | 72 | |
XM_011534064.2 | XP_011532366.1 | 6 - 7 | 828 | 72 | |
XM_011534065.2 | XP_011532367.1 | 5 - 6 | 538 | 72 | |
XM_011534066.2 | XP_011532368.1 | 4 - 5 | 1341 | 72 | |
XM_017007118.1 | XP_016862607.1 | 5 - 6 | 807 | 72 | |
XM_017007119.1 | XP_016862608.1 | 5 - 6 | 467 | 72 | |
XM_017007120.1 | XP_016862609.1 | 3 - 4 | 475 | 72 | |
GenBank mRNA | AB307708.1 | - | 2 - 3 | 271 | 72 |
AK290590.1 | - | 3 - 4 | 633 | 72 | |
BC051670.1 | - | 4 - 5 | 463 | 72 | |
BF307171.1 | - | 3 - 4 | 289 | 72 | |
HQ995528.1 | - | 3 - 4 | 328 | 72 | |
L27586.1 | - | 4 - 5 | 550 | 72 |