The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039492.2 | NP_001034581.1 | 6 - 7 | 911 | 104 |
NM_001318894.1 | NP_001305823.1 | 5 - 6 | 1157 | 104 | |
NM_001318895.1 | NP_001305824.1 | 6 - 7 | 1145 | 104 | |
NM_001318896.1 | NP_001305825.1 | 5 - 6 | 860 | 104 | |
NM_001318897.1 | NP_001305826.1 | 4 - 5 | 911 | 104 | |
NM_001318898.1 | NP_001305827.1 | 5 - 6 | 970 | 104 | |
NM_001318899.1 | NP_001305828.1 | 7 - 8 | 1033 | 104 | |
NM_001450.3 | NP_001441.4 | 5 - 6 | 1094 | 104 | |
NM_201555.1 | NP_963849.1 | 7 - 8 | 1266 | 104 | |
NM_201557.3 | NP_963851.2 | 5 - 6 | 1086 | 104 | |
XM_011510798.2 | XP_011509100.1 | 6 - 7 | 893 | 104 | |
GenBank mRNA | AB158503.1 | - | 4 - 5 | 633 | 104 |
AK313841.1 | - | 6 - 7 | 943 | 104 | |
AL521653.3 | - | 5 - 6 | 787 | 104 | |
AY634685.1 | - | 4 - 5 | 687 | 104 | |
BC012742.2 | - | 7 - 8 | 1266 | 104 | |
BC014397.1 | - | 7 - 8 | 1266 | 104 | |
BC036085.1 | - | 5 - 6 | 741 | 104 | |
BC093049.1 | - | 5 - 6 | 852 | 104 | |
BI092736.1 | - | 2 - 3 | 204 | 104 | |
BT006960.1 | - | 4 - 5 | 687 | 104 | |
CR457437.1 | - | 4 - 5 | 687 | 104 | |
CR936682.1 | - | 5 - 6 | 1157 | 104 | |
CR936799.1 | - | 5 - 6 | 6138 | 104 | |
DQ266432.1 | - | 6 - 7 | 1145 | 104 | |
DQ307067.1 | - | 5 - 6 | 1024 | 104 | |
JQ313902.1 | - | 4 - 5 | 657 | 104 | |
L42176.1 | - | 6 - 7 | 825 | 104 | |
U29332.1 | - | 5 - 6 | 808 | 104 | |
U60117.1 | - | 2 - 3 | 310 | 104 |