The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002880.3 | NP_002871.1 | 4 - 5 | 838 | 60 |
XM_005265355.2 | XP_005265412.1 | 4 - 5 | 566 | 60 | |
XM_005265357.2 | XP_005265414.1 | 4 - 5 | 754 | 60 | |
XM_005265358.4 | XP_005265415.1 | 3 - 4 | 720 | 60 | |
XM_005265359.4 | XP_005265416.1 | 3 - 4 | 720 | 60 | |
XM_011533974.2 | XP_011532276.1 | 5 - 6 | 881 | 60 | |
XM_011533975.2 | XP_011532277.1 | 4 - 5 | 770 | 60 | |
XM_017006966.1 | XP_016862455.1 | 4 - 5 | 542 | 60 | |
GenBank mRNA | AK226028.1 | - | 4 - 5 | 754 | 60 |
AK303561.1 | - | 3 - 4 | 641 | 60 | |
AK312248.1 | - | 4 - 5 | 732 | 60 | |
BC018119.2 | - | 4 - 5 | 752 | 60 | |
KU178297.1 | - | 3 - 4 | 423 | 60 | |
KU178298.1 | - | 3 - 4 | 423 | 60 | |
KU178299.1 | - | 3 - 4 | 423 | 60 | |
X03484.1 | - | 4 - 5 | 552 | 60 |