The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002880.3 | NP_002871.1 | 6 - 7 | 1096 | 66 |
XM_005265355.2 | XP_005265412.1 | 6 - 7 | 824 | 66 | |
XM_005265358.4 | XP_005265415.1 | 5 - 6 | 978 | 66 | |
XM_011533974.2 | XP_011532276.1 | 7 - 8 | 1139 | 66 | |
XM_011533975.2 | XP_011532277.1 | 6 - 7 | 1028 | 66 | |
GenBank mRNA | AK226028.1 | - | 6 - 7 | 1012 | 66 |
AK303561.1 | - | 5 - 6 | 899 | 66 | |
AK312248.1 | - | 6 - 7 | 990 | 66 | |
BC018119.2 | - | 6 - 7 | 1010 | 66 | |
KU178297.1 | - | 5 - 6 | 681 | 66 | |
KU178298.1 | - | 5 - 6 | 681 | 66 | |
X03484.1 | - | 6 - 7 | 810 | 66 |