Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSEN2
Entrez Gene ID
80746
Gene Name
tRNA splicing endonuclease subunit 2
Gene Aliases
PCH2B, SEN2, SEN2L
Chromosome Location
Chr.3: 12484432 - 12539624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145392.1 NP_001138864.1 3 - 4 408 106
NM_001145393.2 NP_001138865.1 3 - 4 376 106
NM_001145394.1 NP_001138866.1 3 - 4 376 106
NM_001321277.1 NP_001308206.1 3 - 4 376 106
NM_001321278.1 NP_001308207.1 3 - 4 376 106
NM_001321279.1 NP_001308208.1 3 - 4 642 106
NM_025265.3 NP_079541.1 3 - 4 660 106
NR_135607.1 3 - 4 376 106
XM_017007292.1 XP_016862781.1 3 - 4 328 163
XM_017007293.1 XP_016862782.1 3 - 4 660 163
XM_017007294.1 XP_016862783.1 3 - 4 394 163
XM_017007295.1 XP_016862784.1 3 - 4 590 163
XM_017007296.1 XP_016862785.1 3 - 4 956 106
XM_017007297.1 XP_016862786.1 3 - 4 660 163
GenBank mRNA AK027821.1 - 3 - 4 660 106
AK074794.1 - 3 - 4 307 106
AK225875.1 - 3 - 4 365 106
AK291183.1 - 3 - 4 445 106
AK300449.1 - 3 - 4 625 106
BC004178.2 - 3 - 4 393 106
BC004211.2 - 3 - 4 393 106
BC019582.1 - 3 - 4 642 106
CN341716.1 - 2 - 3 113 106
DA614108.1 - 3 - 4 377 106

View Full Product Details