The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297748.1 | NP_001284677.1 | 4 - 5 | 696 | 75 |
NM_001297749.1 | NP_001284678.1 | 3 - 4 | 519 | 75 | |
NM_001297750.1 | NP_001284679.1 | 6 - 7 | 893 | 75 | |
NM_001297751.1 | NP_001284680.1 | 5 - 6 | 837 | 75 | |
NM_016511.3 | NP_057595.2 | 5 - 6 | 795 | 75 | |
XM_011520687.2 | XP_011518989.1 | 6 - 7 | 876 | 75 | |
GenBank mRNA | AF200949.1 | - | 5 - 6 | 733 | 75 |
AK075114.1 | - | 5 - 6 | 715 | 75 | |
AK091989.1 | - | 4 - 5 | 647 | 75 | |
AK291584.1 | - | 5 - 6 | 683 | 75 | |
AK300979.1 | - | 3 - 4 | 406 | 75 | |
AK313589.1 | - | 5 - 6 | 713 | 75 | |
AY358587.1 | - | 5 - 6 | 795 | 75 | |
BC039072.1 | - | 5 - 6 | 701 | 75 | |
BC067746.1 | - | 5 - 6 | 715 | 75 | |
BQ722745.1 | - | 5 - 6 | 724 | 75 | |
BX339075.2 | - | 5 - 6 | 733 | 75 | |
BX380212.2 | - | 6 - 7 | 750 | 75 |