Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF7
Entrez Gene ID
51533
Gene Name
PHD finger protein 7
Gene Aliases
HSPC045, HSPC226, NYD-SP6
Chromosome Location
Chr.3: 52410508 - 52423641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278221.2 NP_001265150.1 1 - 2 736 95
NM_016483.6 NP_057567.3 1 - 2 736 95
XM_011533826.2 XP_011532128.1 1 - 2 686 95
GenBank mRNA AY014283.2 - 1 - 2 310 95
BC022002.1 - 1 - 2 595 95

View Full Product Details