The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321126.1 | NP_001308055.1 | 9 - 10 | 1566 | 65 |
NM_001321127.1 | NP_001308056.1 | 9 - 10 | 1699 | 65 | |
NM_016483.6 | NP_057567.3 | 9 - 10 | 1607 | 65 | |
GenBank mRNA | AK223531.1 | - | 8 - 9 | 824 | 65 |
AK292221.1 | - | 9 - 10 | 1413 | 65 | |
AY014283.2 | - | 9 - 10 | 1181 | 65 | |
BC022002.1 | - | 9 - 10 | 1466 | 65 |