Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR31
Entrez Gene ID
114987
Gene Name
WD repeat domain 31
Gene Aliases
-
Chromosome Location
Chr.9: 113313222 - 113340340 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006615.2 NP_001006616.1 2 - 3 276 62
NM_001012361.3 NP_001012361.1 2 - 3 276 62
NM_145241.4 NP_660284.1 2 - 3 276 62
NR_134677.1 2 - 3 276 62
XM_011518194.2 XP_011516496.1 2 - 3 262 62
GenBank mRNA AK074891.1 - 2 - 3 190 62
AK093240.1 - 2 - 3 205 62
AK313843.1 - 2 - 3 215 62
AL833635.1 - 2 - 3 193 62
BC012352.1 - 2 - 3 208 62
BC022982.1 - 2 - 3 202 62
BX279630.1 - 2 - 3 232 62
DA209351.1 - 2 - 3 276 62
DA305733.1 - 2 - 3 199 62

View Full Product Details