The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256012.1 | NP_001242941.1 | 20 - 21 | 2485 | 77 |
NM_001256095.1 | NP_001243024.1 | 19 - 20 | 2419 | 77 | |
NM_005964.3 | NP_005955.3 | 18 - 19 | 2392 | 77 | |
XM_011523875.2 | XP_011522177.1 | 20 - 21 | 2774 | 77 | |
XM_011523878.2 | XP_011522180.1 | 19 - 20 | 2763 | 77 | |
XM_011523879.2 | XP_011522181.1 | 19 - 20 | 2708 | 77 | |
XM_011523880.2 | XP_011522182.1 | 18 - 19 | 2681 | 77 | |
XM_017024677.1 | XP_016880166.1 | 19 - 20 | 2477 | 77 | |
XM_017024678.1 | XP_016880167.1 | 20 - 21 | 2459 | 77 | |
XM_017024679.1 | XP_016880168.1 | 20 - 21 | 2500 | 77 | |
XM_017024680.1 | XP_016880169.1 | 20 - 21 | 2808 | 77 | |
XM_017024681.1 | XP_016880170.1 | 19 - 20 | 2429 | 77 | |
XM_017024682.1 | XP_016880171.1 | 19 - 20 | 2404 | 77 | |
GenBank mRNA | AB210026.1 | - | 20 - 21 | 2437 | 77 |
BC008968.2 | - | 2 - 3 | 96 | 77 | |
BC117690.1 | - | 17 - 18 | 2294 | 77 | |
BC117691.1 | - | 17 - 18 | 2294 | 77 | |
BC144668.1 | - | 19 - 20 | 2376 | 77 | |
BC150634.1 | - | 19 - 20 | 2376 | 77 | |
BE799557.1 | - | - | 436 | 77 | |
M69181.1 | - | 18 - 19 | 2332 | 77 |