The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256012.1 | NP_001242941.1 | 39 - 40 | 5528 | 62 |
NM_001256095.1 | NP_001243024.1 | 38 - 39 | 5462 | 62 | |
NM_005964.3 | NP_005955.3 | 37 - 38 | 5435 | 62 | |
XM_011523875.2 | XP_011522177.1 | 39 - 40 | 5817 | 62 | |
XM_011523878.2 | XP_011522180.1 | 38 - 39 | 5806 | 62 | |
XM_011523879.2 | XP_011522181.1 | 38 - 39 | 5751 | 62 | |
XM_011523880.2 | XP_011522182.1 | 37 - 38 | 5724 | 62 | |
XM_017024677.1 | XP_016880166.1 | 38 - 39 | 5520 | 62 | |
XM_017024678.1 | XP_016880167.1 | 39 - 40 | 5502 | 62 | |
XM_017024679.1 | XP_016880168.1 | 39 - 40 | 5543 | 62 | |
XM_017024680.1 | XP_016880169.1 | 39 - 40 | 5851 | 62 | |
XM_017024681.1 | XP_016880170.1 | 38 - 39 | 5472 | 62 | |
XM_017024682.1 | XP_016880171.1 | 38 - 39 | 5447 | 62 | |
GenBank mRNA | AB210026.1 | - | 39 - 40 | 5480 | 62 |
AK026977.1 | - | 12 - 13 | 1982 | 62 | |
BC000280.1 | - | 6 - 7 | 783 | 62 | |
BC117690.1 | - | 36 - 37 | 5337 | 62 | |
BC117691.1 | - | 36 - 37 | 5337 | 62 | |
BC144668.1 | - | 38 - 39 | 5419 | 62 | |
BC150634.1 | - | 38 - 39 | 5419 | 62 | |
M69181.1 | - | 37 - 38 | 5375 | 62 | |
S67247.1 | - | 1 - 2 | 47 | 62 |