Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASCC2
Entrez Gene ID
84164
Gene Name
activating signal cointegrator 1 complex subunit 2
Gene Aliases
ASC1p100, p100
Chromosome Location
Chr.22: 29788608 - 29838304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242906.1 NP_001229835.1 13 - 14 1478 139
NM_032204.4 NP_115580.2 14 - 15 1706 139
XM_005261775.2 XP_005261832.1 15 - 16 1746 139
XM_011530442.2 XP_011528744.1 15 - 16 1746 139
XM_011530443.2 XP_011528745.1 16 - 17 1812 139
XM_011530444.2 XP_011528746.1 16 - 17 1859 139
XM_011530445.2 XP_011528747.1 15 - 16 1793 139
XM_011530446.2 XP_011528748.1 14 - 15 1676 139
XM_011530448.2 XP_011528750.1 14 - 15 1579 139
XM_011530449.2 XP_011528751.1 13 - 14 1517 139
XM_011530450.2 XP_011528752.1 16 - 17 1862 139
XM_011530451.2 XP_011528753.1 17 - 18 1936 139
XM_011530452.2 XP_011528754.1 16 - 17 1909 139
XM_011530453.2 XP_011528755.1 15 - 16 1764 139
XM_011530454.2 XP_011528756.1 14 - 15 1648 139
XM_011530455.2 XP_011528757.1 13 - 14 1577 139
XM_017028991.1 XP_016884480.1 14 - 15 3884 139
XM_017028992.1 XP_016884481.1 14 - 15 3884 139
XM_017028993.1 XP_016884482.1 13 - 14 3809 139
XM_017028994.1 XP_016884483.1 13 - 14 3809 139
XM_017028995.1 XP_016884484.1 13 - 14 3725 139
XM_017028996.1 XP_016884485.1 15 - 16 2686 139
XM_017028997.1 XP_016884486.1 13 - 14 3725 139
XM_017028998.1 XP_016884487.1 15 - 16 2686 139
XM_017028999.1 XP_016884488.1 16 - 17 1812 139
XM_017029000.1 XP_016884489.1 15 - 16 1793 139
XM_017029001.1 XP_016884490.1 14 - 15 1579 139
XM_017029002.1 XP_016884491.1 13 - 14 1517 139
XM_017029003.1 XP_016884492.1 15 - 16 1653 139
XM_017029004.1 XP_016884493.1 12 - 13 1442 139
XM_017029005.1 XP_016884494.1 16 - 17 1862 139
XM_017029006.1 XP_016884495.1 17 - 18 1936 139
XM_017029007.1 XP_016884496.1 15 - 16 1764 139
XM_017029008.1 XP_016884497.1 14 - 15 1648 139
XM_017029009.1 XP_016884498.1 13 - 14 1577 139
GenBank mRNA AK025241.1 - 6 - 7 735 139
AK222723.1 - 6 - 7 735 139
AK223033.1 - 6 - 7 734 139
AK303257.1 - 13 - 14 1448 139
AL096741.1 - 2 - 3 396 139
AY013289.1 - 13 - 14 1583 139
BC025368.2 - 14 - 15 1667 139

View Full Product Details