Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASCC2
Entrez Gene ID
84164
Gene Name
activating signal cointegrator 1 complex subunit 2
Gene Aliases
ASC1p100, p100
Chromosome Location
Chr.22: 29788608 - 29838304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242906.1 NP_001229835.1 17 - 18 1940 69
NM_032204.4 NP_115580.2 18 - 19 2168 69
XM_005261775.2 XP_005261832.1 19 - 20 2208 69
XM_017028992.1 XP_016884481.1 18 - 19 4346 69
XM_017028994.1 XP_016884483.1 17 - 18 4271 69
XM_017028997.1 XP_016884486.1 17 - 18 4187 69
XM_017028998.1 XP_016884487.1 19 - 20 3148 69
XM_017028999.1 XP_016884488.1 20 - 21 2274 69
XM_017029000.1 XP_016884489.1 19 - 20 2255 69
XM_017029001.1 XP_016884490.1 18 - 19 2041 69
XM_017029002.1 XP_016884491.1 17 - 18 1979 69
XM_017029003.1 XP_016884492.1 19 - 20 2115 69
XM_017029004.1 XP_016884493.1 16 - 17 1904 69
XM_017029005.1 XP_016884494.1 20 - 21 2324 69
XM_017029006.1 XP_016884495.1 21 - 22 2398 69
XM_017029007.1 XP_016884496.1 19 - 20 2226 69
XM_017029008.1 XP_016884497.1 18 - 19 2110 69
XM_017029009.1 XP_016884498.1 17 - 18 2039 69
GenBank mRNA AK022886.1 - 20 - 21 2165 69
AK025241.1 - 10 - 11 1197 69
AK222723.1 - 10 - 11 1197 69
AK303257.1 - 17 - 18 1910 69
AL096741.1 - 5 - 6 944 69
AY013289.1 - 17 - 18 2045 69
BC025368.2 - 18 - 19 2129 69
BU508026.1 - 5 - 6 518 69

View Full Product Details