The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005308.2 | NP_005299.1 | 4 - 5 | 571 | 86 |
XM_005269707.1 | XP_005269764.1 | 4 - 5 | 568 | 86 | |
XM_005269708.1 | XP_005269765.1 | 3 - 4 | 461 | 86 | |
GenBank mRNA | AK298261.1 | - | 4 - 5 | 514 | 86 |
AK301450.1 | - | 3 - 4 | 262 | 86 | |
AK309761.1 | - | 5 - 6 | 692 | 86 | |
AK313012.1 | - | 4 - 5 | 418 | 86 | |
BC064506.1 | - | 4 - 5 | 565 | 86 | |
L15388.1 | - | 4 - 5 | 558 | 86 |