The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005308.2 | NP_005299.1 | 5 - 6 | 669 | 68 |
XM_005269707.1 | XP_005269764.1 | 5 - 6 | 666 | 68 | |
XM_005269708.1 | XP_005269765.1 | 4 - 5 | 559 | 68 | |
GenBank mRNA | AK298261.1 | - | 5 - 6 | 612 | 68 |
AK301450.1 | - | 4 - 5 | 360 | 68 | |
AK309761.1 | - | 6 - 7 | 790 | 68 | |
AK313012.1 | - | 5 - 6 | 516 | 68 | |
BC064506.1 | - | 5 - 6 | 663 | 68 | |
L15388.1 | - | 5 - 6 | 656 | 68 |