The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005308.2 | NP_005299.1 | 8 - 9 | 969 | 66 |
XM_005269707.1 | XP_005269764.1 | 8 - 9 | 966 | 66 | |
XM_005269708.1 | XP_005269765.1 | 7 - 8 | 859 | 66 | |
GenBank mRNA | AK298261.1 | - | 8 - 9 | 912 | 66 |
AK301450.1 | - | 7 - 8 | 660 | 66 | |
AK309761.1 | - | 9 - 10 | 1090 | 66 | |
AK313012.1 | - | 8 - 9 | 816 | 66 | |
BC064506.1 | - | 8 - 9 | 963 | 66 | |
L15388.1 | - | 8 - 9 | 956 | 66 |